Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166400.1 | NP_001159872.1 | 4 | 625 | ||
NM_001166401.1 | NP_001159873.1 | 3 | 597 | |||
NM_005364.4 | NP_005355.2 | 3 | 346 | |||
XM_017029520.1 | XP_016885009.1 | 4 | 413 | |||
GenBank mRNA | AK313943.1 | - | 3 | 346 | ||
BC002455.2 | - | 3 | 292 | 3345962 | ||
BC012744.2 | - | 3 | 285 | 3617812 | ||
BC098120.1 | - | 3 | 221 | |||
BI092675.1 | - | 3 | 518 | |||
BT007340.1 | - | 1 | 76 |