Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005908.3 | NP_005899.3 | 5 | 707 | ||
XM_017008203.1 | XP_016863692.1 | 6 | 693 | |||
GenBank mRNA | AB209846.1 | - | 5 | 654 | ||
AK291598.1 | - | 5 | 695 | |||
AK300015.1 | - | 4 | 477 | |||
AK312827.1 | - | 5 | 662 | |||
AU132611.1 | - | 5 | 707 | |||
BC015743.1 | - | 5 | 697 | 4905535 | ||
EU009130.1 | - | 5 | 607 | |||
U60337.1 | - | 5 | 694 |