Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000898.4 | NP_000889.3 | 6 | 766 | ||
XM_005272608.3 | XP_005272665.1 | 8 | 866 | |||
XM_017029523.1 | XP_016885012.1 | 8 | 916 | |||
XM_017029524.1 | XP_016885013.1 | 7 | 726 | |||
GenBank mRNA | AK294306.1 | - | 7 | 684 | ||
AK298942.1 | - | 6 | 703 | |||
AK312679.1 | - | 6 | 741 | |||
BC022494.1 | - | 6 | 712 | 4800208 | ||
M69177.1 | - | 6 | 667 | |||
S62734.1 | - | 6 | 590 |