Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002373.5 | NP_002364.5 | 4 | 6703 | ||
XM_017022189.1 | XP_016877678.1 | 5 | 6993 | |||
GenBank mRNA | U38292.1 | - | 1 | 6236 |
Gene Symbol: KRTAP6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC104426.1 | - | 1 | 122 | ||
BC104427.1 | - | 1 | 115 |