Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MATN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002379.3 | NP_002370.1 | 5 | 892 | ||
GenBank mRNA | AK075519.1 | - | 5 | 895 | ||
AK312949.1 | - | 5 | 892 | |||
BC131710.1 | - | 5 | 810 |