Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002401.3 | NP_002392.2 | 10 | 1109 | ||
NM_203351.1 | NP_976226.1 | 11 | 1202 | |||
XM_005257376.3 | XP_005257433.1 | 11 | 1422 | |||
XM_005257377.3 | XP_005257434.1 | 10 | 821 | |||
XM_005257378.2 | XP_005257435.1 | 11 | 1434 | |||
GenBank mRNA | AK225784.1 | - | 11 | 1206 | ||
AK308152.1 | - | 5 | 318 | |||
AK315305.1 | - | 11 | 888 | |||
AL834303.1 | - | 11 | 1201 | |||
AX775739.1 | - | 12 | 949 | |||
AX775741.1 | - | 12 | 949 | |||
BC008336.1 | - | 9 | 694 | |||
BC090859.1 | - | 10 | 1110 | 6183752 | ||
BC093672.1 | - | 10 | 840 | 7939517 | ||
BC093674.1 | - | 10 | 840 | 7939519 | ||
BC143224.1 | - | 10 | 828 | |||
BC143235.1 | - | 10 | 837 | |||
DQ454151.1 | - | 9 | 778 | |||
U78876.1 | - | 10 | 873 |