Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002401.3 | NP_002392.2 | 4 | 511 | ||
NM_203351.1 | NP_976226.1 | 5 | 604 | |||
XM_005257376.3 | XP_005257433.1 | 5 | 836 | |||
XM_005257377.3 | XP_005257434.1 | 4 | 235 | |||
XM_005257378.2 | XP_005257435.1 | 5 | 836 | |||
GenBank mRNA | AK225784.1 | - | 5 | 620 | ||
AK315305.1 | - | 5 | 290 | |||
AL834303.1 | - | 5 | 603 | |||
AX775739.1 | - | 6 | 351 | |||
AX775741.1 | - | 6 | 351 | |||
BC008336.1 | - | 3 | 96 | |||
BC090859.1 | - | 4 | 512 | 6183752 | ||
BC093672.1 | - | 4 | 242 | 7939517 | ||
BC093674.1 | - | 4 | 242 | 7939519 | ||
BC143224.1 | - | 4 | 242 | |||
BC143235.1 | - | 4 | 239 | |||
BM479726.1 | - | 604 | ||||
BQ643432.1 | - | 512 | ||||
U78876.1 | - | 4 | 275 |