Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTAGE5 SIRNA Assays ›
Gene Symbol:
CTAGE5
Gene Name:
CTAGE family member 5, ER export factor
Gene Aliases:
MEA6, MGEA, MGEA11, MGEA6
Chromosome Location:
Chr.14: 39265264 - 39351193 on Build GRCh38
UniGene ID:
Hs.509200
Species:
Human
siRNA ID
HSS106515
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTAGE5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001247988.1
NP_001234917.1
22
2224
NM_001247989.1
NP_001234918.1
23
2537
NM_001247990.1
NP_001234919.1
23
2427
NM_005930.3
NP_005921.2
23
2522
NM_203354.2
NP_976229.1
23
2273
NM_203355.2
NP_976230.1
22
2393
NM_203356.2
NP_976231.1
23
2353
XM_005267648.4
XP_005267705.1
23
2233
XM_006720148.2
XP_006720211.1
23
2235
XM_011536775.2
XP_011535077.1
24
2600
XM_011536778.2
XP_011535080.1
24
2288
XM_011536784.2
XP_011535086.1
23
2151
XM_011536785.2
XP_011535087.1
23
2143
XM_017021314.1
XP_016876803.1
24
2616
XM_017021315.1
XP_016876804.1
24
2317
XM_017021316.1
XP_016876805.1
24
2304
XM_017021317.1
XP_016876806.1
24
2725
XM_017021318.1
XP_016876807.1
24
2346
XM_017021319.1
XP_016876808.1
24
2282
XM_017021320.1
XP_016876809.1
23
2308
XM_017021321.1
XP_016876810.1
23
2472
XM_017021322.1
XP_016876811.1
22
2152
XM_017021323.1
XP_016876812.1
24
2100
XM_017021324.1
XP_016876813.1
24
2188
XM_017021325.1
XP_016876814.1
22
2112
XM_017021326.1
XP_016876815.1
22
2022
XM_017021327.1
XP_016876816.1
22
2014
XM_017021328.1
XP_016876817.1
22
2015
XM_017021329.1
XP_016876818.1
22
1994
XM_017021330.1
XP_016876819.1
24
2225
GenBank mRNA
AB209528.1
-
22
2831
AF338233.1
-
23
2367
AF338234.1
-
23
2400
AK091252.1
-
22
2602
AK315254.1
-
22
2114
AL832233.1
-
22
2214
AL833415.1
-
21
1975
BC039017.1
-
23
2315
BC051363.1
-
23
2140
4576721
BC064355.1
-
23
2483
4803802
BX640994.1
-
23
2255
BX648014.1
-
23
2255
U73682.1
-
22
2103
U94780.1
-
23
2500
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
ER to Golgi vesicle-mediated transport
protein exit from endoplasmic reticulum
cargo loading into vesicle
positive regulation of catalytic activity
protein localization to endoplasmic reticulum exit site
regulation of guanyl-nucleotide exchange factor activity
protein binding
enzyme activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0