Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145354.1 | NP_001138826.1 | 3, 4 | 310 | ||
NM_001321316.1 | NP_001308245.1 | 2, 3 | 238 | |||
NM_013255.4 | NP_037387.2 | 2, 3 | 238 | |||
XM_006715993.2 | XP_006716056.1 | 2, 3 | 238 | |||
XM_011516224.2 | XP_011514526.1 | 2, 3 | 238 | |||
GenBank mRNA | AF047489.1 | - | 2, 3 | 192 | ||
AK002024.1 | - | 2, 3 | 177 | |||
AK294384.1 | - | 3, 4 | 311 | |||
AK310452.1 | - | 2, 3 | 191 | |||
AL705061.1 | - | 2, 3 | 182 | |||
BC067825.1 | - | 2, 3 | 195 | 5268939 | ||
BU945366.1 | - | 199 | ||||
DA172589.1 | - | 2, 3 | 238 |