Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002446.3 | NP_002437.2 | 3 | 1231 | ||
XM_011526981.2 | XP_011525283.1 | 3 | 1318 | |||
XM_011526982.2 | XP_011525284.1 | 3 | 1317 | |||
GenBank mRNA | X90846.1 | - | 3 | 1231 | ||
Z48615.1 | - | 3 | 1103 |