Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002423.4 | NP_002414.1 | 2 | 290 | ||
GenBank mRNA | AK293359.1 | - | 2 | 284 | ||
AK309687.1 | - | 2 | 287 | |||
BP332706.1 | - | 2 | 284 | |||
CB109660.1 | - | 2 | 202 | |||
X07819.1 | - | 2 | 264 | |||
Z11887.1 | - | 2 | 264 |