Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYB SIRNA Assays ›
Gene Symbol:
MYB
Gene Name:
MYB proto-oncogene, transcription factor
Gene Aliases:
Cmyb, c-myb, c-myb_CDS, efg
Chromosome Location:
Chr.6: 135180981 - 135219173 on Build GRCh38
UniGene ID:
Hs.606320
Species:
Human
siRNA ID
HSS106821
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130172.1
NP_001123644.1
7
987
NM_001130173.1
NP_001123645.1
7
987
NM_001161656.1
NP_001155128.1
7
987
NM_001161657.1
NP_001155129.1
7
987
NM_001161658.1
NP_001155130.1
7
987
NM_001161659.1
NP_001155131.1
7
987
NM_001161660.1
NP_001155132.1
7
987
NM_005375.3
NP_005366.2
7
987
NR_134958.1
7
987
NR_134959.1
7
987
NR_134960.1
7
987
NR_134961.1
7
987
NR_134962.1
6
766
NR_134963.1
7
987
NR_134964.1
7
987
NR_134965.1
7
987
GenBank mRNA
AF104863.1
-
7
865
AJ606317.1
-
7
987
AJ606318.1
-
7
987
AJ606319.1
-
7
987
AJ606320.1
-
7
987
AJ606321.1
-
7
987
AJ606322.1
-
7
987
AJ606323.1
-
7
987
AJ606324.1
-
7
987
AJ616235.1
-
7
902
AJ616797.1
-
1
26
AY787443.1
-
7
788
AY787444.1
-
5
495
AY787445.1
-
6
716
AY787446.1
-
6
567
AY787447.1
-
7
788
AY787448.1
-
7
788
AY787449.1
-
7
788
AY787450.1
-
7
788
AY787451.1
-
7
788
AY787452.1
-
7
788
AY787453.1
-
7
788
AY787454.1
-
7
788
AY787455.1
-
7
788
AY787456.1
-
7
788
AY787457.1
-
7
788
AY787458.1
-
7
788
AY787459.1
-
7
788
AY787460.1
-
7
788
AY787461.1
-
7
788
AY787462.1
-
7
788
AY787463.1
-
7
788
AY787464.1
-
7
788
AY787465.1
-
7
788
AY787466.1
-
7
788
AY787467.1
-
7
788
AY787468.1
-
7
788
AY787469.1
-
7
788
AY787470.1
-
7
788
AY787471.1
-
7
788
AY787472.1
-
7
788
AY787473.1
-
7
788
AY787474.1
-
7
788
AY787475.1
-
7
788
BC035996.1
-
1
27
BC064955.1
-
7
939
6069320
M13665.1
-
5
650
M13666.1
-
5
650
M15024.1
-
7
901
X52125.1
-
7
985
X52126.1
-
6
791
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
mitotic cell cycle
chromatin remodeling
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of T-helper cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
positive regulation of histone H3-K9 methylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0