Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080779.1 | NP_001074248.1 | 19 | 2218 | ||
NM_001080950.1 | NP_001074419.1 | 19 | 1987 | |||
NM_033375.4 | NP_203693.3 | 19 | 2002 | |||
GenBank mRNA | AB210015.1 | - | 19 | 2168 | ||
AK292910.1 | - | 19 | 2197 | |||
AK295761.1 | - | 19 | 2157 | |||
AK304782.1 | - | 19 | 2045 | |||
BC044891.1 | - | 18 | 1899 | 4470566 | ||
BC068013.1 | - | 19 | 2212 | 6144867 | ||
BC127784.1 | - | 5 | 381 | |||
X98507.1 | - | 18 | 1958 |