Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012334.2 | NP_036466.2 | 34 | 5032 | ||
XM_005248306.4 | XP_005248363.1 | 16 | 3926 | |||
XM_005248307.2 | XP_005248364.1 | 16 | 3402 | |||
XM_006714475.2 | XP_006714538.1 | 33 | 4739 | |||
XM_011514046.2 | XP_011512348.1 | 16 | 3105 | |||
GenBank mRNA | AB018342.2 | - | 34 | 5019 | ||
AF132021.1 | - | 34 | 4644 | |||
AF234532.1 | - | 34 | 4786 | |||
AF247457.2 | - | 34 | 4717 | |||
AK074717.1 | - | 10 | 1378 | |||
AK295630.1 | - | 9 | 1280 | |||
AK295736.1 | - | 16 | 2887 | |||
AK303459.1 | - | 4 | 626 | |||
AK316138.1 | - | 16 | 3060 | |||
AL832428.1 | - | 24 | 3486 | |||
BC137168.1 | - | 34 | 4741 | 9021165 | ||
BC150285.1 | - | 34 | 5019 | 8860110 |