Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAGLU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000263.3 | NP_000254.2 | 3 | 1000 | ||
XM_006721920.2 | XP_006721983.1 | 3 | 393 | |||
XM_011524840.1 | XP_011523142.1 | 3 | 393 | |||
XM_017024686.1 | XP_016880175.1 | 2 | 533 | |||
XM_017024687.1 | XP_016880176.1 | 2 | 737 | |||
GenBank mRNA | BC053991.1 | - | 3 | 683 | 6213852 | |
L78464.1 | - | 3 | 666 | |||
U40846.1 | - | 3 | 761 | |||
U43573.1 | - | 3 | 660 |