Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195193.1 | NP_001182122.1 | 9 | 1965 | ||
NM_002482.3 | NP_002473.2 | 11 | 2157 | |||
NM_152298.3 | NP_689511.2 | 10 | 1140 | |||
XM_005270888.2 | XP_005270945.1 | 12 | 2177 | |||
XM_005270889.2 | XP_005270946.1 | 11 | 1160 | |||
XM_011541509.1 | XP_011539811.1 | 12 | 2177 | |||
XM_017001356.1 | XP_016856845.1 | 10 | 1087 | |||
GenBank mRNA | AK092829.1 | - | 5 | 3178 | ||
AK131249.1 | - | 6 | 2272 | |||
AK222778.1 | - | 11 | 2100 | |||
AK222816.1 | - | 11 | 2100 | |||
AK291637.1 | - | 11 | 2100 | |||
AK298893.1 | - | 9 | 1908 | |||
AK299584.1 | - | 10 | 1964 | |||
BC003113.1 | - | 10 | 1058 | 3504220 | ||
BC010105.2 | - | 11 | 2085 | 3538429 | ||
BC011580.1 | - | 11 | 2083 | 4558913 | ||
BC053608.1 | - | 10 | 1063 | 6013263 | ||
BT006757.1 | - | 10 | 981 | |||
M97856.1 | - | 11 | 2080 |