Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004548.2 | NP_004539.1 | 2 | 346 | ||
GenBank mRNA | AF044954.1 | - | 2 | 310 | ||
AF067169.1 | - | 2 | 299 | |||
AF088991.1 | - | 2 | 341 | |||
AK291876.1 | - | 2 | 302 | |||
AK311744.1 | - | 2 | 322 | |||
BC005829.2 | - | 2 | 304 | 2966710 | ||
BC007509.2 | - | 2 | 301 | 3030207 | ||
BP319067.1 | - | 2 | 354 |