Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146099.1 | NP_001139571.1 | 11 | 1266 | ||
NM_002498.2 | NP_002489.1 | 12 | 1317 | |||
NM_152720.2 | NP_689933.1 | 12 | 1336 | |||
NR_027415.2 | 12 | 1316 | ||||
GenBank mRNA | AB072828.1 | - | 10 | 935 | ||
AK131359.1 | - | 10 | 2184 | |||
AK290259.1 | - | 12 | 1049 | |||
AL833951.1 | - | 11 | 4699 | |||
BC019916.1 | - | 12 | 1044 | 5000918 | ||
DA407862.1 | - | 6 | 471 | |||
Z29067.1 | - | 10 | 800 |