Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NUCB2 SIRNA Assays ›
Gene Symbol:
NUCB2
Gene Name:
nucleobindin 2
Gene Aliases:
HEL-S-109, NEFA
Chromosome Location:
Chr.11: 17260340 - 17331881 on Build GRCh38
UniGene ID:
Hs.654599
Species:
Human
siRNA ID
HSS107364
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NUCB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005013.2
NP_005004.1
7
753
XM_017017804.1
XP_016873293.1
9
988
XM_017017805.1
XP_016873294.1
8
904
XM_017017806.1
XP_016873295.1
8
847
XM_017017807.1
XP_016873296.1
8
858
XM_017017808.1
XP_016873297.1
7
792
XM_017017809.1
XP_016873298.1
7
789
XM_017017810.1
XP_016873299.1
9
1259
XM_017017811.1
XP_016873300.1
9
1168
XM_017017812.1
XP_016873301.1
7
753
XM_017017813.1
XP_016873302.1
8
852
XM_017017814.1
XP_016873303.1
8
845
XM_017017815.1
XP_016873304.1
10
1252
XM_017017816.1
XP_016873305.1
8
1022
XM_017017817.1
XP_016873306.1
8
947
XM_017017818.1
XP_016873307.1
6
598
XM_017017819.1
XP_016873308.1
6
543
XM_017017820.1
XP_016873309.1
9
988
XM_017017821.1
XP_016873310.1
9
984
XM_017017822.1
XP_016873311.1
7
756
XM_017017823.1
XP_016873312.1
8
860
XM_017017824.1
XP_016873313.1
8
858
XM_017017825.1
XP_016873314.1
7
776
XM_017017826.1
XP_016873315.1
7
789
XM_017017827.1
XP_016873316.1
9
1215
XM_017017828.1
XP_016873317.1
8
845
XM_017017829.1
XP_016873318.1
6
621
GenBank mRNA
AB231311.1
-
5
508
AF052642.1
-
5
508
AF052643.1
-
5
508
AF052644.1
-
5
508
AF450266.1
-
7
722
AK093209.1
-
13
1749
AK097398.1
-
9
1169
AK128739.1
-
11
2420
AK291507.1
-
7
783
FJ224327.1
-
7
727
KM197170.1
-
3
153
X76732.1
-
7
727
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Gene Ontology Categories:
Process(es)
DNA binding
calcium ion binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0