Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ODF2 SIRNA Assays ›
Gene Symbol:
ODF2
Gene Name:
outer dense fiber of sperm tails 2
Gene Aliases:
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location:
Chr.9: 128455155 - 128501292 on Build GRCh38
UniGene ID:
Hs.129055
Species:
Human
siRNA ID
HSS107414
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ODF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242352.1
NP_001229281.1
13
1716
NM_001242353.1
NP_001229282.1
13
1802
NM_001242354.1
NP_001229283.1
12
1488
NM_002540.4
NP_002531.3
15
2004
NM_153432.1
NP_702910.1
12
1562
NM_153433.1
NP_702911.1
13
1729
NM_153435.1
NP_702913.1
13
1622
NM_153436.1
NP_702914.1
12
1386
NM_153437.2
NP_702915.1
13
1652
NM_153439.1
NP_702917.1
12
1505
NM_153440.1
NP_702918.1
12
1432
XM_005252010.2
XP_005252067.1
12
1514
XM_005252011.2
XP_005252068.1
12
1454
XM_005252012.3
XP_005252069.1
13
1704
XM_005252013.3
XP_005252070.1
13
1651
XM_005252014.3
XP_005252071.1
13
1700
XM_005252017.4
XP_005252074.1
13
1605
XM_005252018.2
XP_005252075.1
14
1810
XM_005252019.3
XP_005252076.1
14
1829
XM_005252022.2
XP_005252079.1
13
1628
XM_005252023.2
XP_005252080.1
13
1599
XM_005252024.3
XP_005252081.1
13
1618
XM_005252026.4
XP_005252083.1
13
1547
XM_005252029.2
XP_005252086.1
13
1628
XM_006717126.3
XP_006717189.1
13
1704
XM_011518722.2
XP_011517024.1
13
1628
XM_011518723.2
XP_011517025.1
13
1571
XM_011518724.2
XP_011517026.1
14
1792
XM_011518725.2
XP_011517027.1
14
1796
XM_011518726.2
XP_011517028.1
14
1814
XM_011518728.2
XP_011517030.1
13
1565
XM_011518729.2
XP_011517031.1
13
1501
XM_011518730.2
XP_011517032.1
13
1628
XM_011518731.2
XP_011517033.1
12
1457
XM_011518732.2
XP_011517034.1
13
1414
XM_011518733.2
XP_011517035.1
12
1400
XM_011518744.1
XP_011517046.1
12
1454
XM_017014764.1
XP_016870253.1
14
1785
XM_017014765.1
XP_016870254.1
14
1804
XM_017014766.1
XP_016870255.1
12
1514
XM_017014767.1
XP_016870256.1
14
1796
XM_017014768.1
XP_016870257.1
13
1588
XM_017014769.1
XP_016870258.1
13
1617
XM_017014770.1
XP_016870259.1
14
1772
XM_017014771.1
XP_016870260.1
14
1753
XM_017014772.1
XP_016870261.1
14
1829
XM_017014773.1
XP_016870262.1
14
1805
XM_017014774.1
XP_016870263.1
13
1515
XM_017014775.1
XP_016870264.1
14
1772
XM_017014776.1
XP_016870265.1
14
1753
XM_017014777.1
XP_016870266.1
13
1555
XM_017014778.1
XP_016870267.1
13
1658
XM_017014779.1
XP_016870268.1
12
1399
XM_017014780.1
XP_016870269.1
13
1582
XM_017014781.1
XP_016870270.1
12
1337
GenBank mRNA
AF012549.1
-
13
1592
AF053970.1
-
11
1291
AK093367.1
-
12
1512
AK299303.1
-
13
1564
AK300793.1
-
12
1337
AK301754.1
-
12
1429
AK301842.1
-
12
1354
AK301855.1
-
12
1408
AK302684.1
-
13
1656
AY319414.1
-
12
1538
AY366499.1
-
13
1499
BC010629.1
-
15
1802
3874501
BC091500.1
-
13
1616
30378678
DQ444713.1
-
11
1329
DQ444714.1
-
11
1272
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
multicellular organism development
spermatid development
structural molecule activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-7dvbj:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0