Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000436.3 | NP_000427.1 | 10 | 1342 | ||
XM_017009494.1 | XP_016864983.1 | 10 | 1223 | |||
GenBank mRNA | AK299668.1 | - | 6 | 530 | ||
AK312327.1 | - | 10 | 1038 | |||
BC009001.1 | - | 10 | 1066 | 4148643 | ||
CR457049.1 | - | 10 | 1011 | |||
U62961.1 | - | 10 | 1109 |