Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAFAH1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000430.3 | NP_000421.1 | 2, 3 | 590 | ||
XM_017024699.1 | XP_016880188.1 | 2, 3 | 340 | |||
XM_017024700.1 | XP_016880189.1 | 2, 3 | 417 | |||
XM_017024701.1 | XP_016880190.1 | 2, 3 | 333 | |||
GenBank mRNA | AF208835.1 | - | 1, 2 | 22 | ||
AF208836.1 | - | 1, 2 | 22 | |||
AF208837.1 | - | 1, 2 | 22 | |||
AF208838.1 | - | 1, 2 | 22 | |||
AF400434.1 | - | 1, 2 | 215 | |||
AK313078.1 | - | 2, 3 | 244 | |||
BC064638.1 | - | 2, 3 | 232 | 5786560 | ||
L13385.1 | - | 2, 3 | 239 | |||
L13386.1 | - | 2, 3 | 475 |