Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280547.1 | NP_001267476.1 | 1, 2 | 497 | ||
NM_001280548.1 | NP_001267477.1 | 1, 2 | 497 | |||
NM_001280549.1 | NP_001267478.1 | 1, 2 | 497 | |||
NM_001280550.1 | NP_001267479.1 | 1, 2 | 497 | |||
NM_001280552.1 | NP_001267481.1 | 1, 2 | 497 | |||
NM_001280553.1 | NP_001267482.1 | 1, 2 | 497 | |||
NM_001280554.1 | NP_001267483.1 | 1, 2 | 497 | |||
NM_001280555.1 | NP_001267484.1 | 1, 2 | 497 | |||
NM_016734.2 | NP_057953.1 | 1, 2 | 497 | |||
NR_103999.1 | 1, 2 | 497 | ||||
NR_104000.1 | 1, 2 | 497 | ||||
XM_011517896.2 | XP_011516198.1 | 1, 2 | 1967 | |||
GenBank mRNA | AY463952.1 | - | 1, 2 | 59 | ||
AY463953.1 | - | 1, 2 | 59 | |||
AY463954.1 | - | 1, 2 | 59 | |||
AY463955.1 | - | 1, 2 | 59 | |||
AY463956.1 | - | 1, 2 | 59 | |||
AY463957.1 | - | 1, 2 | 59 | |||
FJ626421.1 | - | 1, 2 | 49 | |||
FJ626422.1 | - | 1, 2 | 49 | |||
FJ626423.1 | - | 1, 2 | 49 | |||
FJ626424.1 | - | 1, 2 | 49 | |||
FJ626425.1 | - | 1, 2 | 49 | |||
M96944.1 | - | 1, 2 | 124 |