Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000282.3 | NP_000273.2 | 7 | 639 | ||
NM_001127692.2 | NP_001121164.1 | 6 | 561 | |||
NM_001178004.1 | NP_001171475.1 | 7 | 639 | |||
XM_005254059.2 | XP_005254116.1 | 7 | 634 | |||
XM_011521093.1 | XP_011519395.1 | 7 | 634 | |||
XM_017020605.1 | XP_016876094.1 | 7 | 634 | |||
XM_017020606.1 | XP_016876095.1 | 6 | 485 | |||
XM_017020607.1 | XP_016876096.1 | 7 | 450 | |||
XM_017020608.1 | XP_016876097.1 | 7 | 634 | |||
XM_017020609.1 | XP_016876098.1 | 7 | 547 | |||
XM_017020610.1 | XP_016876099.1 | 5 | 901 | |||
XM_017020611.1 | XP_016876100.1 | 7 | 634 | |||
XM_017020612.1 | XP_016876101.1 | 7 | 634 | |||
XM_017020613.1 | XP_016876102.1 | 7 | 634 | |||
XM_017020614.1 | XP_016876103.1 | 6 | 485 | |||
XM_017020615.1 | XP_016876104.1 | 7 | 634 | |||
XM_017020616.1 | XP_016876105.1 | 7 | 634 | |||
XM_017020617.1 | XP_016876106.1 | 4 | 297 | |||
XM_017020618.1 | XP_016876107.1 | 4 | 297 | |||
GenBank mRNA | AF385926.1 | - | 7 | 571 | ||
AK225288.1 | - | 7 | 539 | |||
AK225299.1 | - | 7 | 537 | |||
AK296771.1 | - | 6 | 459 | |||
AK298318.1 | - | 7 | 561 | |||
AK315503.1 | - | 7 | 537 | |||
BC000140.2 | - | 7 | 511 | 3048998 | ||
DA781564.1 | - | 6 | 459 | |||
X14608.1 | - | 7 | 506 | |||
Z36837.1 | - | 3 | 233 |