Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037339.2 | NP_001032416.1 | 4 | 864 | ||
NM_001037340.2 | NP_001032417.1 | 9 | 1344 | |||
NM_001037341.1 | NP_001032418.1 | 11 | 1289 | |||
NM_001297440.1 | NP_001284369.1 | 10 | 1050 | |||
NM_001297441.1 | NP_001284370.1 | 10 | 943 | |||
NM_001297442.1 | NP_001284371.1 | 4 | 868 | |||
NM_002600.3 | NP_002591.2 | 11 | 1244 | |||
XM_005270924.3 | XP_005270981.1 | 5 | 535 | |||
XM_005270925.2 | XP_005270982.1 | 5 | 576 | |||
XM_006710680.3 | XP_006710743.1 | 5 | 601 | |||
XM_011541566.1 | XP_011539868.1 | 9 | 1107 | |||
XM_017001445.1 | XP_016856934.1 | 8 | 696 | |||
XM_017001446.1 | XP_016856935.1 | 8 | 1294 | |||
GenBank mRNA | AB209081.1 | - | 9 | 1253 | ||
AK289969.1 | - | 11 | 1244 | |||
AK290006.1 | - | 9 | 1225 | |||
AK290206.1 | - | 4 | 859 | |||
AK294441.1 | - | 4 | 559 | |||
BC101480.1 | - | 11 | 1184 | 8068986 | ||
BC105040.1 | - | 11 | 1184 | 8144043 | ||
EF595686.1 | - | 4 | 399 | |||
KJ534916.1 | - | 10 | 1098 | |||
KJ534917.1 | - | 9 | 873 | |||
KJ535082.1 | - | 9 | 859 | |||
L12686.1 | - | 4 | 776 | |||
L20966.1 | - | 11 | 1236 | |||
M97515.1 | - | 4 | 863 | |||
U85048.1 | - | 9 | 1053 |