Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000923.4 | NP_000914.2 | 12 | 1550 | ||
NM_001098818.2 | NP_001092288.1 | 11 | 1275 | |||
NM_001098819.2 | NP_001092289.1 | 11 | 1328 | |||
NR_040546.1 | 10 | 1225 | ||||
XM_011528056.2 | XP_011526358.1 | 11 | 1281 | |||
XM_011528058.2 | XP_011526360.1 | 9 | 857 | |||
XM_017026866.1 | XP_016882355.1 | 12 | 1559 | |||
GenBank mRNA | AK295050.1 | - | 11 | 1791 | ||
AK300888.1 | - | 10 | 1175 | |||
AK300983.1 | - | 11 | 1238 | |||
BC109067.1 | - | 7 | 773 | 40034056 | ||
BC109068.2 | - | 8 | 823 | |||
U66346.1 | - | 12 | 1550 | |||
U66347.1 | - | 9 | 2306 | |||
U66348.1 | - | 6 | 731 | |||
U66349.1 | - | 5 | 456 | |||
U88712.1 | - | 11 | 1328 | |||
U88713.1 | - | 11 | 1275 | |||
Z46632.1 | - | 11 | 1422 |