Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006204.3 | NP_006195.3 | 12 | 1649 | ||
XM_017016322.1 | XP_016871811.1 | 10 | 1013 | |||
GenBank mRNA | BG206143.1 | - | 2 | 102 | ||
U31973.1 | - | 12 | 1649 |