Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243137.1 | NP_001230066.1 | 10 | 965 | ||
NM_002605.2 | NP_002596.1 | 10 | 1167 | |||
NM_173454.1 | NP_775656.1 | 9 | 1029 | |||
XM_011521668.2 | XP_011519970.1 | 3 | 1301 | |||
XM_017022308.1 | XP_016877797.1 | 7 | 1398 | |||
XM_017022309.1 | XP_016877798.1 | 8 | 765 | |||
XM_017022310.1 | XP_016877799.1 | 10 | 1704 | |||
XM_017022311.1 | XP_016877800.1 | 11 | 1902 | |||
GenBank mRNA | AF056490.1 | - | 8 | 630 | ||
AF332653.1 | - | 10 | 1114 | |||
AF388183.1 | - | 10 | 1160 | |||
AF388184.1 | - | 9 | 1022 | |||
AF388185.1 | - | 8 | 943 | |||
AF388186.1 | - | 9 | 1081 | |||
AF388187.1 | - | 11 | 1358 | |||
AK074280.1 | - | 10 | 965 | |||
AK127232.1 | - | 7 | 2448 | |||
BC060762.1 | - | 11 | 1230 | 30344110 | ||
BC075822.1 | - | 10 | 1058 | 6296489 |