Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000925.3 | NP_000916.2 | 8, 9 | 843 | ||
NM_001173468.1 | NP_001166939.1 | 9, 10 | 789 | |||
NM_001315536.1 | NP_001302465.1 | 7, 8 | 789 | |||
NR_033384.1 | 7, 8 | 913 | ||||
GenBank mRNA | AB209241.1 | - | 4, 5 | 3533 | ||
AK225569.1 | - | 8, 9 | 818 | |||
AK225621.1 | - | 8, 9 | 823 | |||
AK293153.1 | - | 9, 10 | 755 | |||
AK311514.1 | - | 9, 10 | 694 | |||
AK313022.1 | - | 8, 9 | 818 | |||
AL117618.1 | - | 7, 8 | 754 | |||
BC000439.2 | - | 8, 9 | 810 | 2820672 | ||
BC001924.1 | - | 8, 9 | 810 | 2820672 | ||
BM971427.1 | - | 2 | 698 | |||
CR541911.1 | - | 8, 9 | 800 | |||
J03576.1 | - | 8, 9 | 818 | |||
M34055.1 | - | 8, 9 | 805 | |||
M34056.1 | - | 7, 8 | 728 | |||
M34479.1 | - | 8, 9 | 800 | |||
M54788.1 | - | 8, 9 | 805 | |||
X57778.1 | - | 8, 9 | 819 |