Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000466.2 | NP_000457.1 | 11 | 1932 | ||
NM_001282677.1 | NP_001269606.1 | 11 | 1932 | |||
NM_001282678.1 | NP_001269607.1 | 11 | 1967 | |||
XM_017012319.1 | XP_016867808.1 | 6 | 852 | |||
GenBank mRNA | AB008112.1 | - | 11 | 1903 | ||
AB052090.1 | - | 11 | 1836 | |||
AB052091.1 | - | 11 | 1836 | |||
AB052092.1 | - | 11 | 1836 | |||
AB052093.1 | - | 11 | 1836 | |||
AB052094.1 | - | 11 | 1836 | |||
AB209355.1 | - | 1 | 1512 | |||
AF026086.1 | - | 11 | 1896 | |||
AF030356.1 | - | 11 | 1840 | |||
AK292955.1 | - | 11 | 1837 | |||
AK293757.1 | - | 11 | 1875 | |||
AK293848.1 | - | 6 | 654 | |||
AK295686.1 | - | 8 | 876 | |||
BC035575.1 | - | 11 | 1874 | 5498274 |