Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000466.2 | NP_000457.1 | 19, 20 | 3124 | ||
NM_001282677.1 | NP_001269606.1 | 18, 19 | 2953 | |||
NM_001282678.1 | NP_001269607.1 | 19, 20 | 3159 | |||
XM_017012319.1 | XP_016867808.1 | 14, 15 | 2044 | |||
GenBank mRNA | AB008112.1 | - | 19, 20 | 3095 | ||
AB052090.1 | - | 18, 19 | 2857 | |||
AB052091.1 | - | 19, 20 | 3028 | |||
AB052092.1 | - | 19, 20 | 3028 | |||
AB052093.1 | - | 19, 20 | 3028 | |||
AB052094.1 | - | 19, 20 | 3028 | |||
AF026086.1 | - | 19, 20 | 3088 | |||
AF030356.1 | - | 19, 20 | 3032 | |||
AK292955.1 | - | 19, 20 | 3029 | |||
AK293757.1 | - | 19, 20 | 3067 | |||
AK295686.1 | - | 16, 17 | 2068 | |||
BC035575.1 | - | 19, 20 | 3066 | 5498274 |