Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFKFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271804.1 | NP_001258733.1 | 10 | 1077 | ||
NM_001271805.1 | NP_001258734.1 | 9 | 978 | |||
NM_002625.3 | NP_002616.2 | 10 | 1143 | |||
NR_073450.1 | 9 | 1049 | ||||
XM_017029576.1 | XP_016885065.1 | 10 | 1547 | |||
XM_017029577.1 | XP_016885066.1 | 10 | 1547 | |||
XM_017029578.1 | XP_016885067.1 | 11 | 1629 | |||
GenBank mRNA | AK300724.1 | - | 9 | 978 | ||
AK314089.1 | - | 10 | 1359 | |||
BC096077.1 | - | 6 | 555 | 40000710 | ||
BC096078.2 | - | 10 | 1145 | 40000711 | ||
BC096079.3 | - | 10 | 1144 | 40000713 | ||
JQ425694.1 | - | 9 | 993 | |||
JQ425695.1 | - | 10 | 1021 | |||
X52638.1 | - | 10 | 1151 |