Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002630.3 | NP_002621.1 | 7, 8 | 971 | ||
GenBank mRNA | BC073740.1 | - | 7, 8 | 953 | 6296399 | |
J04443.1 | - | 7, 8 | 911 | |||
U75272.1 | - | 7, 8 | 954 |