Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHKG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258459.1 | NP_001245388.1 | 11 | 1241 | ||
NM_001258460.1 | NP_001245389.1 | 10 | 1118 | |||
NM_006213.4 | NP_006204.1 | 10 | 1145 | |||
NR_047689.1 | 9 | 1090 | ||||
XM_005271772.4 | XP_005271829.1 | 8 | 1362 | |||
XM_017012324.1 | XP_016867813.1 | 11 | 1280 | |||
XM_017012325.1 | XP_016867814.1 | 8 | 941 | |||
XM_017012326.1 | XP_016867815.1 | 9 | 1500 | |||
XM_017012327.1 | XP_016867816.1 | 8 | 909 | |||
GenBank mRNA | AK293180.1 | - | 11 | 1196 | ||
AK299429.1 | - | 9 | 1046 | |||
AK300928.1 | - | 10 | 1070 | |||
AK312851.1 | - | 10 | 1145 | |||
BC051327.1 | - | 10 | 1083 | 6277230 | ||
BC069655.1 | - | 10 | 1070 | 7262499 | ||
BC069679.1 | - | 10 | 1061 | 7262511 | ||
BC069738.1 | - | 10 | 1062 | 7262523 | ||
BC069754.1 | - | 10 | 1062 | 7262535 | ||
BC074753.2 | - | 9 | 989 | 30915212 | ||
X80590.1 | - | 10 | 1069 |