Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195291.2 | NP_001182220.2 | 7 | 1435 | ||
NM_001271822.1 | NP_001258751.1 | 6 | 860 | |||
NM_001271823.1 | NP_001258752.1 | 6 | 958 | |||
NM_001271824.1 | NP_001258753.1 | 6 | 1342 | |||
NM_001271825.1 | NP_001258754.1 | 6 | 944 | |||
NM_001297699.1 | NP_001284628.1 | 6 | 1585 | |||
NM_001297700.1 | NP_001284629.1 | 6 | 1241 | |||
NM_004568.5 | NP_004559.4 | 6 | 1300 | |||
XM_011514672.1 | XP_011512974.1 | 8 | 1238 | |||
XM_011514673.1 | XP_011512975.1 | 6 | 1392 | |||
XM_011514674.2 | XP_011512976.1 | 6 | 977 | |||
XM_011514676.2 | XP_011512978.1 | 6 | 787 | |||
XM_017010940.1 | XP_016866429.1 | 7 | 799 | |||
XM_017010941.1 | XP_016866430.1 | 5 | 2428 | |||
GenBank mRNA | AB209563.1 | - | 5 | 968 | ||
AF116727.1 | - | 3 | 1521 | |||
AK027149.1 | - | 5 | 2126 | |||
AK057138.1 | - | 5 | 2659 | |||
AK094983.1 | - | 7 | 1435 | |||
AK098566.1 | - | 6 | 957 | |||
AK122895.1 | - | 4 | 1563 | |||
AK122961.1 | - | 5 | 1576 | |||
AK128637.1 | - | 4 | 2989 | |||
AK314578.1 | - | 6 | 1019 | |||
BC001394.2 | - | 6 | 882 | 3051381 | ||
BC098564.1 | - | 6 | 738 | 30563105 | ||
BX538343.1 | - | 6 | 5012 | |||
Z22658.1 | - | 6 | 731 |