Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136528.1 | NP_001130000.1 | 4 | 764 | ||
NM_001136530.1 | NP_001130002.1 | 4 | 694 | |||
NM_006216.3 | NP_006207.1 | 4 | 764 | |||
NR_073116.1 | 5 | 1182 | ||||
XM_005246641.2 | XP_005246698.1 | 4 | 853 | |||
XM_017004329.1 | XP_016859818.1 | 4 | 745 | |||
XM_017004330.1 | XP_016859819.1 | 4 | 582 | |||
XM_017004331.1 | XP_016859820.1 | 4 | 745 | |||
XM_017004332.1 | XP_016859821.1 | 4 | 582 | |||
GenBank mRNA | A03911.1 | - | 3 | 545 | ||
AK225823.1 | - | 4 | 732 | |||
AK295564.1 | - | 4 | 662 | |||
AK297588.1 | - | 4 | 569 | |||
BC015663.2 | - | 4 | 721 | 4578406 | ||
BC042628.1 | - | 5 | 1181 | 4824856 | ||
M17783.1 | - | 3 | 521 |