Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243186.1 | NP_001230115.1 | 4 | 700 | ||
NM_002648.3 | NP_002639.1 | 4 | 700 | |||
GenBank mRNA | AK301158.1 | - | 4 | 258 | ||
BC020224.1 | - | 4 | 633 | 4591723 | ||
DQ022562.1 | - | 4 | 769 | |||
M16750.1 | - | 4 | 292 | |||
M24779.1 | - | 4 | 558 | |||
M54915.1 | - | 4 | 620 |