Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145311.1 | NP_001138783.1 | 2 | 129 | ||
NM_002666.4 | NP_002657.3 | 2 | 200 | |||
XM_005254934.4 | XP_005254991.1 | 2 | 58 | |||
GenBank mRNA | AB005293.1 | - | 2 | 143 | ||
AK310907.1 | - | 2 | 129 | |||
BC031084.1 | - | 2 | 143 | 5284402 | ||
CB266707.1 | - | 2 | 139 |