Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025179.3 | NP_079455.3 | 25 | 5355 | ||
XM_005273164.2 | XP_005273221.1 | 25 | 5419 | |||
XM_005273165.3 | XP_005273222.1 | 25 | 5419 | |||
GenBank mRNA | AB007932.1 | - | 25 | 4806 | ||
BC006193.2 | - | 3 | 270 | |||
BC009343.2 | - | 8 | 1345 | |||
BC132676.1 | - | 25 | 4710 | 40146698 | ||
BC136530.1 | - | 25 | 4710 | 9020520 | ||
X87831.2 | - | 23 | 3260 |