Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198713.1 | NP_001185642.1 | 2 | 356 | ||
NM_001256782.1 | NP_001243711.1 | 3 | 453 | |||
NM_001256822.1 | NP_001243751.1 | 2 | 425 | |||
NM_004574.4 | NP_004565.1 | 2 | 556 | |||
NM_080415.3 | NP_536340.1 | 2 | 327 | |||
NM_080416.3 | NP_536341.1 | 2 | 327 | |||
NR_037155.2 | 2 | 556 | ||||
NR_104196.1 | 2 | 556 | ||||
NR_104197.1 | 2 | 327 | ||||
XM_006721949.2 | XP_006722012.1 | 3 | 385 | |||
GenBank mRNA | AB002110.1 | - | 2 | 359 | ||
AB008753.1 | - | 2 | 391 | |||
AF035811.1 | - | 2 | 392 | |||
AF073312.1 | - | 2 | 387 | |||
AF176379.1 | - | 2 | 293 | |||
AK094579.1 | - | 2 | 1078 | |||
AK126631.1 | - | 1 | 1794 | |||
AK294094.1 | - | 2 | 359 | |||
AK295527.1 | - | 2 | 280 | |||
AK301914.1 | - | 3 | 453 | |||
AK302146.1 | - | 2 | 356 | |||
AK314528.1 | - | 2 | 439 | |||
AK315045.1 | - | 2 | 403 | |||
AK315396.1 | - | 2 | 281 | |||
BC018055.1 | - | 2 | 244 | |||
BC018056.1 | - | 2 | 270 | 4811662 | ||
BC128515.1 | - | 2 | 389 | |||
CR457111.1 | - | 2 | 263 | |||
D89278.1 | - | 2 | 263 | |||
DA076433.1 | - | 2 | 506 | |||
DA343015.1 | - | 2 | 425 | |||
U88829.1 | - | 2 | 388 | |||
U88870.1 | - | 2 | 273 |