Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000939.3 | NP_000930.1 | 3 | 1084 | ||
NM_001035256.2 | NP_001030333.1 | 4 | 1134 | |||
NM_001319204.1 | NP_001306133.1 | 4 | 1164 | |||
NM_001319205.1 | NP_001306134.1 | 3 | 1114 | |||
GenBank mRNA | BC065832.1 | - | 3 | 796 | 30392189 | |
BT019918.1 | - | 2 | 710 | |||
CR541826.1 | - | 2 | 710 | |||
DQ884404.1 | - | 2 | 719 | |||
M38297.1 | - | 2 | 695 |