Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271899.1 | NP_001258828.1 | 7 | 977 | ||
NM_001271900.1 | NP_001258829.1 | 8 | 1171 | |||
NM_001271948.1 | NP_001258877.1 | 7 | 1504 | |||
NM_181674.2 | NP_858060.2 | 7 | 1302 | |||
NM_181675.3 | NP_858061.2 | 7 | 1024 | |||
NM_181676.2 | NP_858062.1 | 6 | 1108 | |||
NM_181677.2 | NP_858063.1 | 8 | 934 | |||
NM_181678.2 | NP_858064.1 | 8 | 947 | |||
NR_073526.1 | 6 | 1419 | ||||
NR_073527.1 | 7 | 1056 | ||||
GenBank mRNA | AK056192.1 | - | 7 | 984 | ||
AK289717.1 | - | 7 | 953 | |||
AK294659.1 | - | 8 | 1080 | |||
AK295347.1 | - | 7 | 948 | |||
AK314810.1 | - | 7 | 1301 | |||
BC031790.1 | - | 7 | 1192 | 4939981 | ||
BX647887.1 | - | 7 | 1038 | |||
M64930.1 | - | 6 | 1230 |