Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282179.1 | NP_001269108.1 | 3 | 869 | ||
NM_001282180.1 | NP_001269109.1 | 3 | 872 | |||
NM_001282181.1 | NP_001269110.1 | 2 | 239 | |||
NM_001282182.1 | NP_001269111.1 | 2 | 194 | |||
NM_006246.3 | NP_006237.1 | 3 | 872 | |||
GenBank mRNA | AB209811.1 | - | 3 | 825 | ||
AB451233.1 | - | 2 | 282 | |||
AB451358.1 | - | 2 | 282 | |||
AK299520.1 | - | 3 | 809 | |||
AK301796.1 | - | 2 | 239 | |||
AK313500.1 | - | 3 | 725 | |||
AK316430.1 | - | 2 | 193 | |||
BC034994.1 | - | 4 | 866 | |||
BC048305.1 | - | 3 | 605 | |||
BC064358.1 | - | 3 | 852 | |||
BC073873.1 | - | 3 | 824 | |||
BC092477.1 | - | 3 | 842 | 5267163 | ||
BC093766.1 | - | 3 | 340 | 7939611 | ||
BC101479.1 | - | 3 | 340 | 8068985 | ||
BC143231.1 | - | 3 | 337 | 9051732 | ||
BC143234.1 | - | 3 | 340 | 9051735 | ||
L76703.1 | - | 3 | 849 | |||
Z69029.1 | - | 3 | 787 |