Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002772.2 | NP_002763.2 | 22 | 2540 | ||
XM_011529654.1 | XP_011527956.1 | 24 | 2814 | |||
XM_011529655.1 | XP_011527957.1 | 25 | 2732 | |||
XM_011529656.2 | XP_011527958.1 | 25 | 2739 | |||
XM_011529657.1 | XP_011527959.1 | 23 | 2769 | |||
XM_011529658.1 | XP_011527960.1 | 22 | 2733 | |||
XM_011529659.1 | XP_011527961.1 | 23 | 2704 | |||
GenBank mRNA | BC111749.1 | - | 22 | 2529 | 40008569 | |
KF688184.1 | - | 2 | 157 | |||
U09860.1 | - | 22 | 2549 |