Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191037.1 | NP_001177966.1 | 20 | 2444 | ||
NM_002807.3 | NP_002798.2 | 20 | 2444 | |||
NR_034059.1 | 19 | 2274 | ||||
GenBank mRNA | AK307415.1 | - | 19 | 2271 | ||
AK307513.1 | - | 18 | 2146 | |||
AK312528.1 | - | 20 | 2401 | |||
BC001053.2 | - | 20 | 2401 | |||
BC014013.2 | - | 20 | 2413 | |||
BC020223.1 | - | 20 | 2391 | |||
BC032445.1 | - | 20 | 2416 | |||
BC039845.1 | - | 20 | 2367 | |||
BC047897.1 | - | 20 | 2366 | |||
BC064398.1 | - | 20 | 2415 | |||
BC073833.1 | - | 20 | 2368 | |||
BC094720.1 | - | 20 | 2430 | 4826340 | ||
BC112344.1 | - | 20 | 2405 | 40008454 | ||
BC114434.1 | - | 20 | 2405 | 40008444 | ||
D44466.1 | - | 20 | 2362 |