Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281528.1 | NP_001268457.1 | 10 | 760 | ||
NM_001281529.1 | NP_001268458.1 | 10 | 721 | |||
NM_006263.3 | NP_006254.1 | 10 | 760 | |||
NM_176783.2 | NP_788955.1 | 10 | 760 | |||
GenBank mRNA | AI445328.1 | - | 1, 2 | 250 | ||
AK312211.1 | - | 10 | 695 | |||
BC000352.2 | - | 10 | 746 | 2961448 | ||
BC007503.2 | - | 10 | 746 | 2961448 | ||
BM470561.1 | - | 10 | 657 | |||
BM981616.1 | - | 2 | 268 | |||
BQ060125.1 | - | 10 | 718 | |||
BT019337.1 | - | 10 | 655 | |||
BU731923.1 | - | 1, 2 | 260 | |||
BX248293.1 | - | 6 | 428 | |||
CR456780.1 | - | 10 | 655 | |||
CR541658.1 | - | 10 | 655 | |||
GQ472237.1 | - | 10 | 746 | |||
L07633.1 | - | 10 | 747 |