Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRD SIRNA Assays ›
Gene Symbol:
PTPRD
Gene Name:
protein tyrosine phosphatase, receptor type D
Gene Aliases:
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location:
Chr.9: 8314246 - 10612840 on Build GRCh38
UniGene ID:
Hs.446083
Species:
Human
siRNA ID
HSS108852
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040712.2
NP_001035802.1
9
1609
NM_001171025.1
NP_001164496.1
8
1588
NM_002839.3
NP_002830.1
21
2258
NM_130391.3
NP_569075.2
10
1618
NM_130392.3
NP_569076.2
10
1618
NM_130393.3
NP_569077.2
9
1600
XM_006716817.3
XP_006716880.1
20
2556
XM_006716823.2
XP_006716886.1
20
2550
XM_006716825.3
XP_006716888.1
20
2552
XM_006716827.3
XP_006716890.1
17
2510
XM_006716832.3
XP_006716895.1
18
2514
XM_006716833.3
XP_006716896.1
20
2546
XM_006716834.3
XP_006716897.1
17
2508
XM_006716835.3
XP_006716898.1
19
2537
XM_006716837.3
XP_006716900.1
19
2526
XM_006716838.3
XP_006716901.1
18
2519
XM_006716839.3
XP_006716902.1
17
2505
XM_011517992.2
XP_011516294.1
20
2549
XM_017014958.1
XP_016870447.1
20
2555
XM_017014959.1
XP_016870448.1
21
2627
XM_017014960.1
XP_016870449.1
19
2516
XM_017014961.1
XP_016870450.1
19
2500
XM_017014962.1
XP_016870451.1
11
1688
XM_017014963.1
XP_016870452.1
20
2555
XM_017014964.1
XP_016870453.1
20
2554
XM_017014965.1
XP_016870454.1
20
2557
XM_017014966.1
XP_016870455.1
19
2546
XM_017014967.1
XP_016870456.1
19
2541
XM_017014968.1
XP_016870457.1
20
2552
XM_017014969.1
XP_016870458.1
19
2537
XM_017014970.1
XP_016870459.1
20
2555
XM_017014971.1
XP_016870460.1
18
2526
XM_017014972.1
XP_016870461.1
20
2555
XM_017014973.1
XP_016870462.1
18
2522
XM_017014974.1
XP_016870463.1
20
2555
XM_017014975.1
XP_016870464.1
17
2511
XM_017014976.1
XP_016870465.1
20
2552
XM_017014977.1
XP_016870466.1
20
2553
XM_017014978.1
XP_016870467.1
20
2552
XM_017014979.1
XP_016870468.1
20
2554
XM_017014980.1
XP_016870469.1
20
2549
XM_017014981.1
XP_016870470.1
17
2505
XM_017014982.1
XP_016870471.1
20
2551
XM_017014983.1
XP_016870472.1
20
2551
XM_017014984.1
XP_016870473.1
19
2542
XM_017014985.1
XP_016870474.1
20
2548
XM_017014986.1
XP_016870475.1
20
2547
XM_017014987.1
XP_016870476.1
19
2529
XM_017014988.1
XP_016870477.1
19
2529
XM_017014989.1
XP_016870478.1
20
2547
XM_017014990.1
XP_016870479.1
20
2546
XM_017014991.1
XP_016870480.1
20
2546
XM_017014992.1
XP_016870481.1
19
2537
XM_017014993.1
XP_016870482.1
20
2544
XM_017014994.1
XP_016870483.1
20
2544
XM_017014995.1
XP_016870484.1
19
2541
GenBank mRNA
AB208869.1
-
8
1435
AB211400.1
-
20
2226
BC106713.1
-
8
1536
40027581
BC106714.1
-
10
1566
40027582
BC106715.2
-
10
1566
40027584
BC106716.1
-
9
1548
40027585
L38929.1
-
11
1668
X54133.1
-
1
348
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
phosphate-containing compound metabolic process
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
transmembrane receptor protein tyrosine phosphatase signaling pathway
chemical synaptic transmission
neuron differentiation
peptidyl-tyrosine dephosphorylation
positive regulation of dendrite morphogenesis
presynaptic membrane assembly
transmembrane receptor protein tyrosine phosphatase activity
receptor binding
protein binding
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0