Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161440.2 | NP_001154912.2 | 5 | 706 | ||
NM_002842.4 | NP_002833.4 | 7 | 1240 | |||
XM_011527183.1 | XP_011525485.1 | 7 | 1261 | |||
XM_011527188.1 | XP_011525490.1 | 6 | 1210 | |||
XM_011527190.1 | XP_011525492.1 | 7 | 1261 | |||
XM_017027056.1 | XP_016882545.1 | 6 | 1248 | |||
XM_017027057.1 | XP_016882546.1 | 6 | 1248 | |||
XM_017027058.1 | XP_016882547.1 | 6 | 1281 | |||
XM_017027059.1 | XP_016882548.1 | 6 | 1248 | |||
XM_017027060.1 | XP_016882549.1 | 6 | 1248 | |||
XM_017027061.1 | XP_016882550.1 | 4 | 714 | |||
XM_017027062.1 | XP_016882551.1 | 6 | 1156 | |||
XM_017027063.1 | XP_016882552.1 | 6 | 1149 | |||
XM_017027064.1 | XP_016882553.1 | 6 | 1248 | |||
GenBank mRNA | AK097257.1 | - | 7 | 1259 | ||
BC111715.1 | - | 6 | 656 | 40008777 | ||
BC111716.1 | - | 7 | 1190 | 40008775 | ||
D15049.2 | - | 7 | 1214 |