Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000318.2 | NP_000309.1 | 4 | 744 | ||
NM_001079867.1 | NP_001073336.1 | 3 | 712 | |||
NM_001172086.1 | NP_001165557.1 | 5 | 750 | |||
NM_001172087.1 | NP_001165558.1 | 3 | 608 | |||
GenBank mRNA | AK312997.1 | - | 2 | 411 | ||
BC000661.2 | - | 4 | 517 | 3347824 | ||
BC005375.1 | - | 3 | 513 | 3934225 | ||
BC093043.1 | - | 4 | 530 | 30399827 | ||
BP193912.1 | - | 3 | 511 | |||
M85038.1 | - | 4 | 551 | |||
M86852.1 | - | 4 | 659 |