Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3IL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013401.3 | NP_037533.2 | 5, 6 | 902 | ||
XM_005274140.4 | XP_005274197.1 | 5, 6 | 867 | |||
XM_006718647.3 | XP_006718710.1 | 5, 6 | 869 | |||
XM_011545196.2 | XP_011543498.1 | 5, 6 | 873 | |||
XM_011545197.2 | XP_011543499.1 | 5, 6 | 1189 | |||
XM_011545198.2 | XP_011543500.1 | 5, 6 | 897 | |||
XM_011545200.2 | XP_011543502.1 | 2, 3 | 308 | |||
GenBank mRNA | AF084557.1 | - | 4, 5 | 456 | ||
BC022239.1 | - | 5, 6 | 818 | 4390573 |